Merck
  • Redox-independent chromium isotope fractionation induced by ligand-promoted dissolution.
Redox-independent chromium isotope fractionation induced by ligand-promoted dissolution.
Nature communications (2017-11-19)
Emily M Saad, Xiangli Wang, Noah J Planavsky, Christopher T Reinhard, Yuanzhi Tang
ABSTRACT
The chromium (Cr) isotope system has emerged as a potential proxy for tracing the Earth's atmospheric evolution based on a redox-dependent framework for Cr mobilization and isotope fractionation. Although studies have demonstrated that redox-independent pathways can also mobilize Cr, no quantitative constraints exist on the associated isotope fractionations. Here we survey the effects of common environmental ligands on the dissolution of Cr(III)-(oxy)hydroxide solids and associated Cr isotope fractionation. For a variety of organic acids and siderophores, δ
Sigma-Aldrich
Enterobactin, from Escherichia coli, ≥98% (HPLC)