Merck
  • Vortex-assisted liquid-liquid-liquid microextraction followed by high performance liquid chromatography for the simultaneous determination of fourteen phenolic acids in honey, iced tea and canned coffee drinks.
Vortex-assisted liquid-liquid-liquid microextraction followed by high performance liquid chromatography for the simultaneous determination of fourteen phenolic acids in honey, iced tea and canned coffee drinks.
Talanta (2017-07-26)
Marwan Shalash, Ahmad Makahleh, Salizawati Muhamad Salhimi, Bahruddin Saad
ABSTRACT
A vortex-assisted liquid-liquid-liquid microextraction method followed by high performance liquid chromatography-diode array detection for the determination of fourteen phenolic acids (cinnamic, m-coumaric, chlorogenic, syringic, ferulic, o-coumaric, p-coumaric, vanillic, p-hydroxybenzoic, caffeic, 2, 4-dihydroxybenzoic, sinapic, gentisic and gallic acids) in honey, iced tea and canned coffee drink samples has been developed. The separation was achieved using a Poroshell 120-EC-C18 column under a gradient elution at a flow rate of 0.6mLmin
Sigma-Aldrich
Propyl acetate, ≥98%, FG
Sigma-Aldrich
Isobutyl acetate, FCC, FG
Sigma-Aldrich
Butyl acetate, JIS special grade, ≥99.0%