- Enantioselective photocycloaddition mediated by chiral Brønsted acids: asymmetric synthesis of the rocaglamides.
Enantioselective photocycloaddition mediated by chiral Brønsted acids: asymmetric synthesis of the rocaglamides.
Journal of the American Chemical Society (2006-06-15)
Baudouin Gerard, Sheharbano Sangji, Daniel J O'Leary, John A Porco
PMID16771486
ABSTRACT
Enantioselective syntheses of methyl rocaglate and the related natural products rocaglamide and rocaglaol are outlined. The approach involves enantioselective [3 + 2] photocycloaddition promoted by chiral Brønsted acids (TADDOLs) to afford an aglain precursor followed by a ketol shift/reduction sequence to the rocaglate core.