Merck
Arl8 and SKIP act together to link lysosomes to kinesin-1.
Developmental cell (2011-12-17)
Cláudia Rosa-Ferreira, Sean Munro
ABSTRACT
Lysosomes move bidirectionally on microtubules, and this motility can be stimulated by overexpression of the small GTPase Arl8. By using affinity chromatography, we find that Arl8-GTP binds to the soluble protein SKIP (SifA and kinesin-interacting protein, aka PLEKHM2). SKIP was originally identified as a target of the Salmonella effector protein SifA and found to bind the light chain of kinesin-1 to activate the motor on the bacteria's replicative vacuole. We show that in uninfected cells both Arl8 and SKIP are required for lysosomes to distribute away from the microtubule-organizing center. We identify two kinesin light chain binding motifs in SKIP that are required for lysosomes to accumulate kinesin-1 and redistribute to the cell periphery. Thus, Arl8 binding to SKIP provides a link from lysosomal membranes to plus-end-directed motility. A splice variant of SKIP that lacks a light chain binding motif does not stimulate movement, suggesting fine-tuning by alternative splicing.
Sigma-Aldrich
Anti-TGN46 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Calreticulin (405-417) Rabbit pAb, liquid, Calbiochem®