Merck
Conversion visual loss: a differential diagnosis in infant amblyopia.
European journal of ophthalmology (2009-11-03)
Carlos Laria, Maria Elena Perez, Eduardo Perez, David Pablo Pinero, José Maria Ruiz-Moreno, Jorge L Aliò
ABSTRACT
To report the first case of amblyopia due to a conversion disorder in a child. A 9-year-old girl without allergies or family or personal antecedents of interest presented to our clinic. She manifested a progressive visual loss after an episode of lumbar pain. This progressive loss led the patient in a 6-month period to legal bilateral blindness: visual acuity inferior to 2/20 in both eyes and severe constriction of both visual fields. Several pathologic processes were discarded by means of multiple explorations and a general multidisciplinary analysis: physical pathology, simulating patient, schizophrenia, and the Briquet syndrome. Finally, she was diagnosed with a conversion somatoform disorder leading to a bilateral blindness. For 6 months, the patient was successfully treated with psychotherapy and a pharmacologic protocol. Total recovery was achieved with final visual acuity of 20/20 in both eyes and normal visual fields. Conversion disorders should be considered as an additional etiology of visual loss in children. In these cases, the analysis and multidisciplinary treatment is crucial for a successful outcome.
Product Description
Supelco
Lormetazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®