Merck
  • RNA aptamers and spiegelmers: synthesis, purification, and post-synthetic PEG conjugation.
RNA aptamers and spiegelmers: synthesis, purification, and post-synthetic PEG conjugation.
Current protocols in nucleic acid chemistry (2011-09-09)
Stefanie Hoffmann, Johannes Hoos, Sven Klussmann, Stefan Vonhoff
ABSTRACT
This unit describes the solid-phase synthesis and downstream processing for RNA oligonucleotides with a length of up to 40 to 50 nucleotides on a 1- to 4-mmol scale with subsequent conjugation to PEG using the L-RNA spiegelmer NOX-E36 as an example. Following synthesis and two-step deprotection, the crude oligonucleotide is purified by preparative reversed-phase HPLC and desalted by tangential flow ultrafiltration. The resulting intermediate amino-modified oligonucleotide is reacted with NHS-ester-activated PEG, and the oligonucleotide-PEG conjugate is obtained after preparative AX-HPLC purification, followed by ultrafiltration and lyophilization. Critical process parameters are described, as well as time considerations and examples for analytical methods used as in-process and quality controls.
Sigma-Aldrich
Activator Reagent 0.5M ETT in Acetonitrile, 0.5M ETT in Acetonitrile