
Ketotifen for peak identification

Synonym(s):
Ketotifen fumarate salt
Empirical Formula (Hill Notation):
C19H19NOS · C4H4O4
CAS Number:
Molecular Weight:
425.50
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing