Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

4.90220

Sigma-Aldrich

Cetylaminhydrofluoride

for synthesis

Synonym(s):

Cetylaminhydrofluoride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H36FN
CAS Number:
Molecular Weight:
261.46
UNSPSC Code:
12352107
EC Index Number:
221-588-7

Quality Level

pH

6.5 (25 °C, 10 g/L in H2O)

mp

122-125 °C

solubility

0.4 g/L

bulk density

230 kg/m3

storage temp.

2-30°C

InChI

1S/C16H35N.FH/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17;/h2-17H2,1H3;1H

InChI key

KMXTYZBQPJXGNK-UHFFFAOYSA-N

Analysis Note

Identity (IR-spectrum): passes test
Assay (HClO₄): 98 - 102 %
Melting range (lower value): ≥ 122 °C
Melting range (upper value): ≤ 125 °C
Fluoride (F): 7.1 - 7.4 %
Methanol: ≤ 0.3 %
Water (K. F.): ≤ 0.5 %

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service