MilliporeSigma
HomePhotometry & ReflectometryDetermination of Nitrite in Compost
Photometric Determination of Nitrite in Compost by the Griess Method Subsequent to Extraction with a Calcium Chloride Solution
Abstract
This protocol outlines the photometric determination of nitrite ion concentration in compost samples by the Griess Method using Spectroquant® test kits. Following an extraction of nitrite ions with a calcium chloride solution, the procedure utilizes a spectrophotometer to measure the nitrite content, ensuring precise results.
Section Overview:
three models of Spectroquant® spectrophotometers: Prove 100 plus, Prove 300 plus, and Prove 600 plus. Each device has a sleek, modern design with a rectangular body. The largest model in the foreground, Prove 600 plus, is green with a black top and features a touchscreen interface displaying measurement data, as well as a slot for sample insertion. The top right corner of the device has a yellow "M" logo and "600+" printed in bold yellow on the front corner. Two smaller models, Prove 100 plus and Prove 300 plus, are placed in the background. Prove 100 plus is light green with a black top, while Prove 300 plus is yellow with a black top. Both also feature touchscreens and the "M" logo.
Spectroquant® spectrophotometers - Prove 100 plus, Prove 300 plus, & Prove 600 plus
Introduction
Nitrite is an anion with the formula NO2- and a rich source of nitrogen found in soil, for plants. In the intermediate step of nitrification, ammonia (NH3) is aerobically oxidized first to nitrites1. Nitrites are found in composts matured to be used to improve crop yield in agricultural fields. It has been studied that nitrites get accumulated in composts after the exudence of ammonia, when only ammonia-oxidizing bacteria get proliferated, with the natural eradication of nitrite oxidizing bacteria (NOB)2. Nitrite accumulation leads to nitrous oxide (N2O), a greenhouse gas3 emitted by soil. It is important to analyze composts for nitrite, to mitigate nitrite accumulation and their role in the consequential impact on the environment. 
This application note details the photometric determination of nitrite in compost using the Griess method, after extraction with a calcium chloride solution. 
Reagents, Instruments and Materials
Nitrite Test Kits & Reagents
For the measurement, one of the following Spectroquant® test kits is necessary:
  • Spectroquant® Nitrite Cell Test (1.14547)
  • Spectroquant® Nitrite Test (1.14776)
Instrument(s) & Devices
For the measurement, one of the following Spectroquant® photometers is necessary:
  • Spectroquant® VIS Spectrophotometer Prove 100 Plus (1.73026)
  • Spectroquant® UV/VIS Spectrophotometer Prove 300 Plus (1.73027)
  • Spectroquant® UV/VIS Spectrophotometer Prove 600 Plus (1.73028)
  • Spectroquant® Colorimeter Move 100 (1.73632)
Also, legacy systems
  • Spectroquant® Spectrophotometer Prove 100/300/600
  • Spectroquant® Photometer NOVA 30/60/60A
Software for Data transfer
  • Optional Spectroquant® Prove Connect to LIMS software package (Y.11086) to transfer your data into an existing LIMS system.
Instrument Accessories
  • Rectangular cells 10 mm (1.14946) and/or
  • Rectangular cells 20 mm (1.14947) and/or
  • Rectangular cells 50 mm (1.14944)
Other Reagents and Accessories
  • Calcium chloride dihydrate for analysis (1.02382)
  • Water for analysis (1.16754)
  • Charcoal activated for soil tests
Experimental Procedure
Reagent Preparation
  • Calcium chloride solution, 0.025 mol/L: Dissolve 3.68 g of calcium chloride dihydrate for analysis with 1 L of water for analysis.
Sample Preparation
  • In a glass bottle mix 50 g of naturally moist sample, free from coarse stones, with 100 mL of a calcium chloride solution 0.025 mol/L.
  • Add a spatula-tip full of charcoal activated for soil tests and shake the closed bottle in a shaking machine for 1 hour (alternative stir in a beaker).
  • Let the suspension settle and filter it through a folded filter.
  • For determination of the water content dry a similar sample to constant weight in the drying kiln at 105 °C.
  • A drying of the sample before the determination is not advisable because of the fast change in the nitrogen forms.
The analysis should take place very quickly. Storing the sample in plastic bags at room temperature changes the analysis values after a short time already
Analysis
Determine with one of the above-mentioned test kits.
Calculation
Nitrate content in mg/kg NO2-= analysis value in mg/L NO2- x 2
See more applications for photometry at Protocols and Application Notes
Related Products
Lo sentimos, se ha producido un error inesperado
Network error: Failed to fetch
References
1.
Beeckman F, Motte H, Beeckman T. 2018. Nitrification in agricultural soils: impact, actors and mitigation. Current Opinion in Biotechnology. 50166-173. https://doi.org/10.1016/j.copbio.2018.01.014
2.
Fukumoto Y, Inubushi K. 2009. Effect of nitrite accumulation on nitrous oxide emission and total nitrogen loss during swine manure composting. Soil Science and Plant Nutrition. 55(3):428-434. https://doi.org/10.1111/j.1747-0765.2009.00376.x
3.
Wilson T, Warren J, Arnal B. 2017. Nitrous oxide emissions from soil - Oklahoma state university. [Internet]. Okstate.edu.[cited 08 Aug 2024]. Available from: https://extension.okstate.edu/fact-sheets/nitrous-oxide-emissions-from-soil