MilliporeSigma
HPLC Analysis of Flavanone Enantiomers on Astec® CYCLOBOND I 2000 DMP, Ethanol:Hexane Mobile Phase
HPLC Analysis of Flavanone Enantiomers on Astec® CYCLOBOND I 2000 DMP, Ethanol:Hexane Mobile Phase application for HPLC
Materiales
analytical column
Referencia del producto
Descripción
Precios
Astec® Cyclobond I 2000 DMP Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
standard
Referencia del producto
Descripción
Precios
Flavanone
98%
CONDITIONS
column
CYCLOBOND I 2000 DMP, 25 cm x 4.6 mm I.D., 5 μm particles (20724AST)
mobile phase
[A] ethanol; [B] hexane; (20:80, A:B)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 254 nm