MilliporeSigma
HPLC Analysis of Ketoprofen Enantiomers on Astec® CHIROBIOTIC® R
HPLC Analysis of Ketoprofen Enantiomers on Astec® CHIROBIOTIC® R application for HPLC
Materiales
Estándar
Referencia del producto
Descripción
Precios
Ketoprofen
meets USP testing specifications
Ketoprofen
≥98% (TLC)
(S)-(+)-Ketoprofen
99%
Ketoprofen
VETRANAL®, analytical standard
columna analítica
Referencia del producto
Descripción
Precios
Astec® Chirobiotic® R Chiral HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
CONDITIONS
column
CHIROBIOTIC R, 15 cm x 4.6 mm I.D., 5 μm particles (13023AST)
mobile phase
[A] 20 mM ammonium acetate, pH 5.6; [B] methanol; (50:50, A:B)
flow rate
0.6 mL/min
column temp.
25 °C
detector
UV, 254 nm