MilliporeSigma
HPLC Analysis of Ribonucleosides on Ascentis® Si
HPLC Analysis of Ribonucleosides on Ascentis® Si application for HPLC
Materiales
Estándar
Referencia del producto
Descripción
Precios
Adenosine
Pharmaceutical Secondary Standard; Certified Reference Material
Uridine
Pharmaceutical Secondary Standard; Certified Reference Material
Guanosine
Pharmaceutical Secondary Standard; Certified Reference Material
Cytidine
certified reference material, pharmaceutical secondary standard
columna analítica
Referencia del producto
Descripción
Precios
Ascentis® Si HPLC Column
5 μm particle size, L × I.D. 10 cm × 2.1 mm
CONDITIONS
column
Ascentis Si, 10 cm x 2.1 mm I.D., 5 μm particles (581508-U)
mobile phase
[A] 150 mM ammonium formate (pH 3.0, with formic acid): [B] acetonitrile; (10:90, A:B)
flow rate
0.2 mL/min
column temp.
35 °C
detector
UV, 270 nm
injection
1 μL
sample
as indicated in mobile phase