MilliporeSigma
Sulfur gases
Sulfur gases suitable for GC, application for SPME
Materiales
Fibra para SPME
Referencia del producto
Descripción
Precios
Fibra de carboxeno/polidimetilsiloxano (DVB/CAR/PDMS) de SPME
df 75 μm(CAR/PDMS, for use with manual holder, needle size 24 ga
columna analítica
Referencia del producto
Descripción
Precios
Columna capilar para GC Supel-Q PLOT
L × I.D. 30 m × 0.32 mm, average thickness 15 μm
CONDITIONS
sample/matrix
sulfur gases in 40 mL vial
SPME fiber
Carboxen/PDMS, 75 μm (57318)
extraction
10 min headspace at 40 °C
desorption process
3 min at 290 °C
column
Supel-Q PLOT, 30 m × 0.32 mm I.D. (24242)
oven
45 °C (1 min) to 250 °C at 15 °C/min
detector
FPD