MilliporeSigma
US EPA Method TO-17: GC Analysis of Volatiles on VOCOL after Collection/Desorption using Carbotrap® 300 Tube
US EPA Method TO-17: GC Analysis of Volatiles on VOCOL after Collection/Desorption using Carbotrap® 300 Tube suitable for GC, application for air monitoring
Materiales
analytical column
Referencia del producto
Descripción
Precios
SPB®-1
L × I.D. 60 m × 0.25 mm, df 3.00 μm
related product
Referencia del producto
Descripción
Precios
Carbotrap® 300
glass TD tube, fritted, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea
standard
Referencia del producto
Descripción
Precios
JHAP-43 Mix
1 ppm in nitrogen, pkg of 110 L (pi-marked cylinder), analytical standard
CONDITIONS
sample/matrix
JHAP-43 Gas Mix (500429), 1ppm each component, 20mL
adsorbent tube
Carbotrap 300 (28283-U)
desorption process
20 mL/min at 330 °C for 6 min (cryo-focused to inlet @ -150°C)
column
SPB-1, 60 m × 0.25 mm I.D., 3.0 μm
oven
35°C (8 min), 5°C/min to 100°C (0 min), 15°C/min to 230°C (8 min)
detector
MSD, scan range 35-269
carrier gas
helium 31cm/sec
Descripción
Legal Information
Carbotrap is a registered trademark of Merck KGaA, Darmstadt, Germany