MilliporeSigma
Oxonitriles: a grignard addition-acylation route to enamides.
Organic letters (2006-10-06)
Fraser F Fleming, Guoqing Wei, Zhiyu Zhang, Omar W Steward
RESUMEN
[reaction: see text] Sequential addition of three different Grignard reagents and pivaloyl chloride to 3-oxo-1-cyclohexene-1-carbonitrile installs four new bonds to generate a diverse array of cyclic enamides. Remarkably, formation of the C-magnesiated nitrile intermediate is followed by preferential acylation by pivaloyl chloride rather than consumption by an in situ Grignard reagent. Rapid N-acylation of the C-magnesiated nitrile generates an acyl ketenimine that reacts readily with Grignard reagents or a trialkylzincate, effectively assembling highly substituted, cyclic enamides.