MilliporeSigma
Re-evaluation of in vitro activity of primycin against prevalent multiresistant bacteria.
International journal of medical microbiology : IJMM (2014-09-03)
Péter Feiszt, Gyula Mestyán, Monika Kerényi, Orsolya Dobay, Judit Szabó, Zsuzsanna Dombrádi, Edit Urbán, Levente Emődy
RESUMEN
With the increasing emergence of antibiotic resistances old antibiotics became a valuable source to find agents suitable to address this problem. More than 20 years after the last report, our purpose was to re-evaluate the in vitro antibacterial activity of the topical agent primycin against current important bacterial pathogens. Minimal inhibitory concentrations (MIC) and minimal bactericidal concentrations (MBC) of primycin were tested in comparison with agents widely applied topically, and with those of mupirocin and vancomycin, the topical and the non-topical gold-standard anti-MRSA agents. Primycin was ineffective (MIC>64 μg/ml) against all the Gram-negative isolates tested. On the other hand, all the tested Gram-positive isolates were susceptible with MIC90 values of 0.06 μg/ml for staphylococci and 0.5-1 μg/ml for enterococci, streptococci, and P. acnes isolates, including all the multiresistant strains. Against MRSA isolates primycin showed slightly higher activity than mupirocin, and inhibited the mupirocin-resistant strains also. MBC90 values ranged from 0.25 to 2 μg/ml for the investigated Gram-positive species. The bactericidal effect proved to be concentration-dependent in time-kill experiments. Spontaneous resistant mutants did not emerge in single-step mutation experiments and the resistance development was very slow by serial passaging. Passaged S. aureus strains showing increased primycin MIC values exhibited elevated vancomycin and daptomycin MIC values also. Though elucidation of the mechanisms behind warrants further investigations, these correlations can be related to development of vancomycin-intermediate phenotype. From the point of view of medical practice it is noteworthy that the increased primycin MIC values remained far below the concentration accessible by local application of the agent. These data make primycin a remarkable object of further investigations as well as a promising candidate for topical application against multiresistant Gram-positive pathogens.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Mupirocin, ≥92% (HPLC), powder
Sigma-Aldrich
Daptomycin, cyclic lipopeptide antibiotic
Sigma-Aldrich
Erythromycin standard solution, 1 mg/mL in H2O
USP
Erythromycin, United States Pharmacopeia (USP) Reference Standard
USP
Mupirocin, United States Pharmacopeia (USP) Reference Standard
USP
Oxytetracycline, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Supelco
Erythromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Oxytetracycline, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
Millipore
Oxytetracycline, suitable for microbiology
Erythromycin, for microbiological assay, European Pharmacopoeia (EP) Reference Standard