MilliporeSigma
Rhodium-catalyzed asymmetric conjugate alkynylation of nitroalkenes.
Chemical communications (Cambridge, England) (2010-08-19)
Takahiro Nishimura, Takahiro Sawano, Sumito Tokuji, Tamio Hayashi
RESUMEN
Asymmetric addition of (triisopropylsilyl)acetylene to nitroalkenes took place in the presence of a rhodium/chiral bisphosphine catalyst to give beta-alkynylated nitroalkanes in high yields with high enantioselectivity.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
(Triisopropylsilyl)acetylene, 97%