MilliporeSigma
Pd-catalyzed oxidative CH/CH direct coupling of heterocyclic N-oxides.
Organic letters (2013-09-12)
Wei Liu, Yahui Li, Yue Wang, Chunxiang Kuang
RESUMEN
A highly efficient protocol for C-H/C-H cross-coupling has been found to occur between 2-aryl-1,2,3-triazole N-oxides and pyridine N-oxide derivatives. In addition, two homocoupling reactions of 2-substituted 1,2,3-triazole N-oxides and some pyridine N-oxide derivatives were developed. A possible pathway of C-H/C-H direct coupling is discussed.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Pyridine N-oxide, 95%