MilliporeSigma
Changes in energy charge and block of protein synthesis in rabbit reticulocytes under the action of valinomycin.
Molecular biology reports (1980-12-31)
H Breitbart, M Herzberg
RESUMEN
Valinomycin blocks protein synthesis at the elongation stage in intact reticulocyte while being without action on a cell free system. We demonstrate here that this membrane mediated inhibition is accompanied by a change of the energy charge (ATP/ADP/AMP ratio) of the cells. There seems to be an accumulation of AMP without a proportional gain in ADP. The change in energy charge in itself can be the cause of the inhibition of protein synthesis. We show also that valinomycin can cause a decrease of the energy charge at concentrations where its action as an ionophore is not seen and independent of a possible activation of the Na-K dependent ATPase.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Dicyclohexano-18-crown-6, 98%