MilliporeSigma
Selective killing of transformed cells by methotrexate with histidine deprivation or with alpha-amino alcohols.
Cancer research (1983-10-01)
E M Newman, D W Nierenberg, D V Santi
RESUMEN
The effects of amino acid deprivation and treatment with amino alcohols upon the growth, viability, and susceptibility to methotrexate (MTX) cytotoxicity were examined in BALB/3T3 cells and their virally transformed counterparts, SV-T2 cells. Cells were deprived of either histidine or tyrosine plus phenylalanine, or they were treated with amino alcohol analogues of histidine and tyrosine (histidinol and tyrosinol). When incubated in medium lacking histidine and supplemented with dialyzed serum (histidine-deficient medium), the BALB/3T3 cells remained viable for at least 3 days, but they ceased logarithmic growth, and the cell number reached an early plateau. In contrast, the SV-T2 cells continued to divide in histidine-deficient medium. Neither cell line ceased division in medium deficient in both phenylalanine and tyrosine. Incubation of the BALB/3T3 cells with 1.5 mM histidinol or 1.0 mM tyrosinol caused an early plateau similar to the effect of histidine deprivation. SV-T2 cells were not affected by these concentrations of histidinol or tyrosinol, but growth was arrested at higher concentrations. Any of the conditions used which caused a plateau in the number of BALB/3T3 cells substantially protected the treated cells from cell death caused by MTX. Therefore, pretreatment of BALB/3T3 cells with histidine deprivation, 1.5 mM histidinol, or 1.0 mM tyrosinol protected this cell line against MTX-induced cell death, while the same pretreatment conditions failed to protect SV-T2 cells. (SV-T2 cells were protected by 4.0 mM histidinol.) Thus, the ability of MTX to kill cells in vitro can be selectively modified by conditions which protect cells which retain normal growth control characteristics, but which do not protect virally transformed cells.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
D-Tyrosinol hydrochloride, 98%