MilliporeSigma
  • Novel polysiloxane resin functionalized with dicyclohexano-18-crown-6 (DCH18C6): Synthesis, characterization and extraction of Sr(II) in high acidity HNO3 medium.
Novel polysiloxane resin functionalized with dicyclohexano-18-crown-6 (DCH18C6): Synthesis, characterization and extraction of Sr(II) in high acidity HNO3 medium.
Journal of hazardous materials (2012-05-23)
Gang Ye, Feifei Bai, Jichao Wei, Jianchen Wang, Jing Chen
RESUMEN
A novel kind of polysiloxane resin functionalized with dicyclohexano-18-crown-6 (DCH18C6) was synthesized through a post-modification approach. The DCH18C6 moieties bearing amino groups were firstly prepared, followed by covalent grafting to a silica precursor P-(CH(2))(3)-Cl (Where P represents a 3-dimentional polymerized silica matrix) based on nucleophilic substitution reaction. (29)Si and (13)C solid-state NMR, FT-IR, XPS, TGA, ESEM and elemental analysis were employed to systematically characterize the structure, thermal property and surface morphology of the functionalized resin. The results indicated that the DCH18C6 ligands were successfully bonded to the polysiloxane resin with a satisfactory grafting degree (33.6wt.%). Due to the robust organosilica framework and the covalent immobilization of the ligands, the functionalized resin had excellent thermal stability and acid resistance. Batch experiments showed that the resin could effectively separate Sr(II) in high acidity mediums. The distribution coefficient (K(d)) of 43.6cm(3)/g could be achieved in 5.0mol/L HNO(3) solution. The influences of contact time and acidity of HNO(3) on the resin's extraction performance were examined. The reusability and the selectivity to Sr(II) over interference ions were investigated. The DCH18C6-functionalized resin might be potentially applied for the radiostrontium removal in the high level liquid waste (HLLW).
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Dicyclohexano-18-crown-6, 98%