MilliporeSigma
Functions of plasmalogen lipids in health and disease.
Biochimica et biophysica acta (2012-05-26)
Nancy E Braverman, Ann B Moser
RESUMEN
Plasmalogens are a unique class of membrane glycerophospholipids containing a fatty alcohol with a vinyl-ether bond at the sn-1 position, and enriched in polyunsaturated fatty acids at the sn-2 position of the glycerol backbone. These two features provide novel properties to these compounds. Although plasmalogens represent up to 20% of the total phospholipid mass in humans their physiological roles have been challenging to identify, and are likely to be particular to different tissues, metabolic processes and developmental stages. Their biosynthesis starts in peroxisomes, and defects at these steps cause the malformation syndrome, Rhizomelic Chondrodysplasia Punctata (RCDP). The RCDP phenotype predicts developmental roles for plasmalogens in bone, brain, lens, lung, kidney and heart. Recent studies have revealed secondary plasmalogen deficiencies associated with more common disorders and allow us to tease out additional pathways dependent on plasmalogen functions. In this review, we present current knowledge of plasmalogen biology in health and disease.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Batyl alcohol, 99%