Merck
Synthesis of phosphonic analogues of carnitine and gamma-amino-beta-hydroxybutyric acid.
Bioorganic chemistry (2004-11-09)
Elzbieta J Tadeusiak
RESUMEN
The involvement of carnitine and gamma-amino-beta-hydroxybutyric acid in the biology of mammalian cells, the physiology of the human body, and some important aspects of medicinal treatment has induced many research groups to develop their pharmacologically potent analogues. Among them are the very important phosphonic analogues: phosphocarnitine and gamma-amino-beta-hydroxypropylphosphonic acid. This mini-review describes the various methodologies used for the synthesis of these compounds.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
4-Amino-3-hydroxybutyric acid, 98%
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
(S)-(+)-4-Amino-3-hydroxybutyric acid, 97%
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad