MilliporeSigma
Glycinebetaine protects the D1/D2/Cytb559 complex of photosystem II against photo-induced and heat-induced inactivation.
Journal of plant physiology (2003-04-11)
Suleyman I Allakhverdiev, Hidenori Hayashi, Yoshitaka Nishiyama, Alexander G Ivanov, Jalal A Aliev, Vyacheslav V Klimov, Norio Murata, Robert Carpentier
RESUMEN
The presence of 1.0 mol/L glycinebetaine during isolation of D1/D2/Cytb559 reaction centre (RC) complexes from photosystem II (PSII) membrane fragments preserved the photochemical activity, monitored as the light-induced reduction of pheophytin and electron transport from diphenylcarbazide to 2.6-dichlorophenol-indophenol.-Glycinebetaine also protected the D1/D2/Cytb559 complexes against strong light-induced damage to the photochemical reactions and the irreversible bleaching of beta-carotene and chlorophyll. The presence of glycinebetaine also enhanced thermotolerance of the D1/D2/Cytb559 complexes isolated in the presence of 1.0 mol/L betaine with an increase in the temperature for 50% inactivation from 29 degrees C to 35 degrees C. The results indicate an increased supramolecular structural stability in the presence of glycinebetaine.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
1,5-Diphenylcarbazide, ACS reagent
Sigma-Aldrich
1,5-Diphenylcarbazide, reagent grade
Supelco
1,5-Diphenylcarbazide, reag. Ph. Eur., ≥98.0%
Sigma-Aldrich
1,5-Diphenylcarbazide, JIS special grade