Merck
Synthesis, intramolecular migrations and enzymic hydrolysis of partially pivaloylated methyl alpha-D-mannopyranosides.
Carbohydrate research (2003-04-02)
Srdanka Tomić, Vesna Petrović, Maja Matanović
RESUMEN
A series of methyl O-pivaloyl-alpha-D-mannopyranosides was synthesized using pivaloyl chloride in pyridine. The 3,6-di-O-pivaloyl derivative 6 undergoes intramolecular transesterification in neutral conditions (buffer, pH 7.2) to give its 2,6-di-O-pivaloyl analogue 5. The course of this migration was followed using 14C-labelled 6. As opposed to 6 compound 5 was shown to be a good substrate for esterases present in rabbit serum. Thus, regioselective enzymic hydrolysis led to the preferential cleavage of the 2-OPiv group to yield a mixture of 2- and 6-O-monopivalates in a ratio of 1:2.6.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Trimethylacetyl chloride, 99%