MilliporeSigma
  • [The degradation of glycoproteins with lithium borohydride. Isolation and analysis of O-glycopeptides with reduced C-terminal amino acid residue].
[The degradation of glycoproteins with lithium borohydride. Isolation and analysis of O-glycopeptides with reduced C-terminal amino acid residue].
Bioorganicheskaia khimiia (2000-05-12)
N P Arbatskiĭ, L M Likhosherstov, M V Serebriakova, O S Brusov, V N Shibaev, V A Derevitskaia, N K Kochetkov
RESUMEN
By the example of fetuin and a blood-group-specific mucin from porcine stomach, we showed that, under conditions of reductive degradation of glycoproteins with LiBH4-LiOH in 70% aqueous tert-butyl alcohol, the reduction and cleavage of amide bonds occur much faster than the simultaneous beta-elimination of carbohydrate chains O-linked with Ser and Thr residues of the peptide chain. The major degradation products containing the O-linked glycans are the O-glycosylated derivatives of 2-aminopropane-1,3-diol and 2-aminobutane-1,3-diol (the products of reduction of glycosylated Ser and Thr) and the glycopeptides containing 2-4 amino acid residues with reduced C-terminal amino acid. Seventeen homogeneous O-glycopeptides were isolated from the fetuin degradation products by ion-exchange and reversed-phase HPLC. Their structures were determined by MALDI-TOF mass spectrometry and by analyses for amino acids, amino alcohols, and carbohydrates. The application of the reaction for characterization of O-glycans and localization of O-glycosylation sites in O- and N,O-glycoproteins is discussed.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Lithium borohydride solution, 2.0 M in THF
Sigma-Aldrich
Lithium borohydride, ≥95.0%
Sigma-Aldrich
Lithium borohydride, ≥90%
Sigma-Aldrich
Lithium borohydride, hydrogen-storage grade, ≥90%
Sigma-Aldrich
Lithium borohydride solution, 0.5 M in diethyl ether