- A turn-on fluorescent indicator for citrate with micromolar sensitivity.
A turn-on fluorescent indicator for citrate with micromolar sensitivity.
Dalton transactions (Cambridge, England : 2003) (2007-09-11)
M Isabel Burguete, Francisco Galindo, Santiago V Luis, Laura Vigara
PMID17828363
RESUMEN
A turn-on fluorescent indicator for citric acid (citrate) has been developed, displaying high emission enhancement (+1500%) and low interference by other carboxylates. The sensor is based on the non-emissive copper(II) complex of a fluorescent amino amide, which, upon addition of citrate decomplexates to yield the emissive ligand. The detection limit estimated for this new chemosensing system is about 0.5 microM. This novel approach to the analysis of citrate constitutes an alternative ca. 10(2)-10(3) times more sensitive than the standard method based on the enzyme citrate lyase.