Merck
  • Roles of diclofenac and its metabolites in immune activation associated with acute hepatotoxicity in TgCYP3A4/hPXR-humanized mice.
Roles of diclofenac and its metabolites in immune activation associated with acute hepatotoxicity in TgCYP3A4/hPXR-humanized mice.
International immunopharmacology (2020-07-03)
Weifan Jiang, Tianming Dai, Shuilin Xie, Lan Ding, Lizhen Huang, Renke Dai
RESUMEN
Diclofenac (DCF) is a widely used nonsteroidal anti-inflammatory drug, but it comes with a high risk of drug-induced liver injury (DILI). Despite the quinone-imine adduct pathways, the immunotoxicity is recently considered as another factor for DILI. However, such immune responses are still elusive. In the present study, investigation of the immune response in the acute hepatotoxicity model of TgCYP3A4/hPXR-humanized mice was conducted by administration of DCF and DCF metabolites, respectively. In a single dose intraperitoneal injection of 80 mg/kg DCF, the pharmacokinetic results showed the major DCF metabolites, including 4'-hydroxy-diclofenac (4'-OH-DCF), 5-hydroxy-diclofenac (5-OH-DCF) and diclofenac glucuronide (DCF-G) were generated after DCF treatment. Not only DCF, but those DCF metabolites could also directly cause different degrees of acute liver injury as significantly increased the serum ALT levels in a short time period in the TgCYP3A4/hPXR-humanized mice. Furthermore, the three DCF metabolites could directly stimulate the significant elevation of serum immune-related factors in varying degrees. Transcriptome analysis revealed the differentially expressed genes in the liver of DCF-G treated mice were mostly involved with the "immune system process" and "cell death" and related to "IL-17 signaling pathway" and "TNF-α signaling pathway", but 5-OH-DCF had little effect on the expressions of those genes. These results indicate that the metabolite DCF-G plays an important role in the activation of the hepatic immune system, which might be involved in the pathogenesis of DCF-induced acute liver injury.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Pentostatin, ≥95% (HPLC)
Supelco
Flufenamic acid, analytical standard, for drug analysis
Supelco
5-Hydroxydiclofenac, analytical standard