MilliporeSigma
The A-kinase anchoring protein 15 regulates feedback inhibition of the epithelial Na+ channel.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2007-01-25)
Abderrahmane Bengrine, Jinqing Li, Mouhamed S Awayda
RESUMEN
Protein kinase A anchoring proteins or AKAPs regulate the activity of many ion channels. Protein kinase A (PKA) is a well-recognized target of AKAPs, with other kinases now emerging as additional targets. We examined the roles of epithelial-expressed AKAPs in regulating the epithelial Na+ channel (ENaC). Experiments used heterologous expression with AKAP15, AKAP-KL, and AKAP79 in Xenopus oocytes. Experiments were carried out under high and low Na+ conditions, as Na+ loading is known to affect the baseline activity of ENaC in a PKC-dependent mechanism. ENaC activity was unaffected by AKAP79 and AKAP-KL expression. However, oocytes coexpressing AKAP15 exhibited an 80% and 91% reduction in the amiloride-sensitive, whole-cell conductance in high and low Na+ conditions, respectively. The reduced channel activity was unaffected by PKA activation or inhibition, indicating a PKA-independent mechanism. Expression with a membrane-targeting domain, mutant form of AKAP15 (AKAP15m) prevented the decrease of ENaC activity, but only under low Na+ conditions. In high sodium conditions, coexpression with AKAP15m led to an increase of ENaC activity to levels similar to those observed under low Na+. These results indicate that membrane-associated AKAP15 reduces ENaC activity whereas the cytoplasmically associated one may participate in the channel's feedback inhibition by intracellular Na+, a process known to involve PKC. This hypothesis was further confirmed in coexpression experiments, which demonstrated functional and physical interaction between AKAP15 and PKCalpha. We propose that AKAP15 regulates ENaC via a novel PKA-independent pathway.