MilliporeSigma
Specific interaction of VEGF165 with beta-amyloid, and its protective effect on beta-amyloid-induced neurotoxicity.
Journal of neurochemistry (2005-03-19)
Seung-Pil Yang, Byoung-Oh Kwon, Yong Song Gho, Chi-Bom Chae
RESUMEN
beta-amyloid (Abeta) is a major component of senile plaques that is commonly found in the brain of Alzheimer's disease (AD) patient. In the previous report, we showed that an important angiogenic factor, vascular endothelial growth factor (VEGF) interacts with Abeta and is accumulated in the senile plaques of AD patients' brains. Here we show that Abeta interacts with VEGF(165) isoform, but not with VEGF(121). Abeta binds to the heparin-binding domain (HBD) of VEGF(165) with similar affinity as that of intact VEGF(165). Abeta binds mostly to the C-terminal subdomain of HBD, but with greatly reduced affinity than HBD. Therefore, the full length of HBD appears to be required for maximal binding of Abeta. Although Abeta binds to heparin-binding sequence of VEGF, it does not bind to other heparin-binding growth factors except midkine. Thus it seems that Abeta recognizes unique structural features of VEGF HBD. VEGF(165) prevents aggregation of Abeta through its HBD. We localized the core VEGF binding site of Abeta at around 26-35 region of the peptide. VEGF(165) and HBD protect PC12 cells from the Abeta-induced cytotoxicity. The mechanism of protection appears to be inhibition of both Abeta-induced formation of reactive oxygen species and Abeta aggregation.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Amyloid β 26-35 human