MilliporeSigma
Highly fluorescent, monolithic semiconductor nanorod clusters for ultrasensitive biodetection.
Chemical communications (Cambridge, England) (2018-09-25)
Yali Yuan, Giulia Adriani, Yang Xu, Yinthai Chan
RESUMEN
We have developed highly fluorescent, monolithic colloidal CdSe seeded CdS nanorod clusters comprising thousands of nanorods. Their use in the sandwich assay detection of a model protein yields a thousand-fold improvement in the detection limit compared to individual nanorods, making them suitable for the detection of low abundance molecular targets.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
1-Biotinyl-3,6-dioxa-8-octaneamine hydrochloride, ≥95%