MilliporeSigma
  • Expression and characterization of two glucuronoyl esterases from Thielavia terrestris and their application in enzymatic hydrolysis of corn bran.
Expression and characterization of two glucuronoyl esterases from Thielavia terrestris and their application in enzymatic hydrolysis of corn bran.
Applied microbiology and biotechnology (2019-02-15)
Jiao Tang, Liangkun Long, Yunfeng Cao, Shaojun Ding
RESUMEN
The thermophilic fungus Thielavia terrestris when cultured on cellulose produces a cocktail of thermal hydrolases with potential application in saccharification of lignocellulosic biomass and other biotechnological areas. Glucuronoyl esterases are considered to play a unique role as accessory enzymes in lignocellulosic material biodegradation by cleaving the covalent ester linkage between 4-O-methyl-D-glucuronic acid (MeGlcA) and lignin in lignin-carbohydrate complexes (LCCs). Two glucuronoyl esterases from T. terrestris named TtGE1 and TtGE2 were expressed in Pichia pastoris. Both esterases displayed features of thermophilic enzymes, with the optimal temperature at 45 °C and 55 °C. TtGE1 and TtGE2 exhibited activity towards methyl (4-nitrophenyl β-D-glucopyranosid) uronate (Me-GlcA-pNP) but no catalytic activity to benzyl-D-glucuronate (BnzGlcA), indicating the difference in substrate specificity from previously studied fungal GEs. A substantial increase in the release of monomeric sugars and glucuronic acid from autohydrolysis of corn bran was observed by the supplementing TtGEs into commercial xylanase; the results clearly demonstrated that the TtGEs played a significant role in this degradation process. This research on TtGEs enriches our knowledge of this novel class of fungal GEs. These newly characterized TtGEs could be used as promising accessory enzymes to improve the hydrolysis efficiency of commercial enzymes in saccharification of lignocellulosic materials due to their thermophilic characteristics.