MilliporeSigma
PECAM-1: a multi-functional molecule in inflammation and vascular biology.
Arteriosclerosis, thrombosis, and vascular biology (2007-09-18)
Abigail Woodfin, Mathieu-Benoit Voisin, Sussan Nourshargh
RESUMEN
Platelet endothelial cell adhesion molecule-1 (PECAM-1 or CD31) is a molecule expressed on all cells within the vascular compartment, being expressed to different degrees on most leukocyte sub-types, platelets, and on endothelial cells where its expression is largely concentrated at junctions between adjacent cells. As well as exhibiting adhesive properties, PECAM-1 is an efficient signaling molecule and is now known to have diverse roles in vascular biology including roles in angiogenesis, platelet function, and thrombosis, mechanosensing of endothelial cell response to fluid shear stress, and regulation of multiple stages of leukocyte migration through venular walls. This review will focus on some new developments with respect to the role of PECAM-1 in inflammation and vascular biology, highlighting the emerging complexities associated with the functions of this unique molecule.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
PECAM-I human, recombinant, expressed in HEK 293 cells, ≥97% (SDS-PAGE), ≥97% (HPLC), suitable for cell culture