EP5820766000
Eppendorf® Rotor Lid
for rotor FA-45-20-17, aerosol-tight
About This Item
Productos recomendados
material
aluminum
description
for 5804/5804R & 5810/5810R
feature
aerosol-tight
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5820766000
technique(s)
centrifugation: suitable
compatibility
for rotor FA-45-20-17
for use with Centrifuge 58xx
General description
Replacement rotor lid, aerosol-tight, for rotor FA-45-20-17
Features and Benefits
aerosol-tight
Legal Information
Eppendorf is a registered trademark of Eppendorf AG