MilliporeSigma
EP0030075420
Eppendorf® BioBased epT.I.P.S.® Reloads
new
volume 0.1—20 μL, sterile, Biopur, L -40 mm, pkg of 480 ea (5 trays x 96 tips)
About This Item
UNSPSC Code:
41106300
sterility
sterile
feature
DNase free
RNase free
graduations
Biopur
packaging
pkg of 480 ea (5 trays x 96 tips)
manufacturer/tradename
Eppendorf® 0030075420
L
-40 mm
volume
0.1—20 μL
suitability
suitable for PCR
suitable for microbiology
suitable for molecular biology
suitable for nucleic acid purification
General description
epT.I.P.S.® BioBased Reloads, Biopur®, 0.1 – 20 µL, 40 mm, medium gray, colorless tips, 480 tips (5 trays × 96 tips)
Application
Molecular Biology, DNA purification, PCR, Microbiology, Cell Biology, Medical Research, Agriculture Research, Environmental Monitoring, Forensic Analysis
Features and Benefits
  • Improve your laboratory carbon footprint and switch to epT.I.P.S. BioBased in the new Eppendorf Reload variants. These pipette tips are produced from second-generation biobased polymer with 90% of the feedstock attributed to renewable resources (i.e. waste and residues from vegetable oilrefining or waste cooking oil), avoiding the use of fossil fuel-based petroleum.
  • Reduce your laboratory waste volume – use Reloads instead of disposable Racks.
  • Eppendorf production facility is ISCC PLUS(International Sustainability & Carbon Certification) certified, confirming the raw materials used at the facility are “bio-circular”. Enables traceabilityalong the entire supply chain.
  • Pipette tips with ACT (Accountability, Consistency,and Transparency) Label - an independent environmental impact factor assessmentthrough non-profit My Green.
  • Material that is both quality and purity certified.
  • Get excellent sample integrity as no slip agents, plasticizers and biocides are used Independent laboratory – Lot-specific purity certificates can be downloaded from Eppendorf website.
  • General quality certificates in compliance with standards and general Eppendorf quality items.
  • Available in volumes from 10 µL to 1,250 µL Long in Reloads for use with epT.I.P.S. Box 2.0.
  • Available as epT.I.P.S. BioBased Biopur, ep Dualfilter T.I.P.S. BioBased PCR clean and sterile, ep Dualfilter T.I.P.S. SealMax BioBased Biopur
Legal Information
Biopur is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG