MilliporeSigma
Todas las fotos(6)
Documentos clave
Z545449
Molymod® Individual organic model set
Amino acid collection set
Seleccione un Tamaño
1 SET
$186.00
$186.00
Fecha estimada de envío14 de abril de 2025
Seleccione un Tamaño
Cambiar Vistas
1 SET
$186.00
About This Item
UNSPSC Code:
60104506
NACRES:
NB.17
$186.00
Fecha estimada de envío14 de abril de 2025
description
models 20
manufacturer/tradename
Molymod
General description
The mushroom-shaped hydrogen Atomlink components and the short (non-visible) bonding links provide compact, semi-space-filling models with an internuclear scale of 1.45 cm per Angstrom. Polymer models include long, flexible links for demonstration of the mechanism of polymerization.
Legal Information
Atomlink is a trademark of Spiring Enterprises Ltd.
Molymod is a registered trademark of Spiring Enterprises Ltd.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)