MilliporeSigma
HPLC Analysis of Ephedrine Enantiomers on CHIRALPAK® AGP
HPLC Analysis of Ephedrine Enantiomers on CHIRALPAK® AGP application for HPLC
Materials
analytical column
Product No.
Description
Pricing
CHIRALPAK® AGP HPLC Column
5 μm particle size, L × I.D. 10 cm × 3 mm
standard
Product No.
Description
Pricing
(1R,2S)-(−)-Ephedrine
98%
DL-Ephedrine hydrochloride
99%
(1R,2S)-(−)-Ephedrine hydrochloride
99%
(1S,2R)-(+)-Ephedrine hydrochloride
99%
CONDITIONS
column
CHIRAL-AGP, 10 cm x 4 mm I.D., 5 μm particles (58170AST)
mobile phase
1 mM octanoic acid in 10 mM (NH4)2HPO4, pH 6.9
flow rate
0.9 mL/min
column temp.
25 °C
detector
UV, 225 nm
injection
10 μL
sample
1 mg/mL in methanol
Description
Legal Information
CHIRALPAK is a registered trademark of Daicel Corp.