MilliporeSigma
HPLC Analysis of Tricyclic Antidepressants in Serum on Discovery® C18 after SPE using Discovery® DSC-18
HPLC Analysis of Tricyclic Antidepressants in Serum on Discovery® C18 after SPE using Discovery® DSC-18 application for SPE, application for HPLC
Materials
Analytical column
Product No.
Description
Pricing
Discovery® C18 HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
related product
Product No.
Description
Pricing
Discovery® DSC-18 SPE Tube
bed wt. 100 mg, volume 1 mL, pk of 108
CONDITIONS
sample preparation
SPE (Solid Phase Extraction)
SPE tube/cartridge
Discovery DSC-18, 100 mg/1 mL (52602-U)
column
Discovery C18, 15 cm × 4.6 mm, 5 μm preceded by a 2 cm C18 guard column and 0.5 μm frit filter (504955)
mobile phase
[A] acetonitrile: [B] methanol: [C] 25 mM potassium phosphate, monobasic (pH 7 with triethlylamine) (45:25:30, A:B:C)
flow rate
1 mL/min
column temp.
ambient
detector
UV, 254 nm
injection
50 μL, diluted porcine serum extract
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany