HPLC Analysis of Oligosaccharides from Dark Corn Syrup on SUPELCOGEL™ Ag
CONDITIONS
column
SUPELCOGEL Ag, 30 cm × 7.8 mm I.D. (59315)
mobile phase
water
flow rate
0.5 mL/min
column temp.
85 °C
detector
RI
injection
10 μL of 1g syrup/10 mL DI water, filtered (0.20 μm filter)
Description
Legal Information
SUPELCOGEL is a trademark of Sigma-Aldrich Co. LLC