MilliporeSigma
HPLC Analysis of Methacrylamide, Acrylamide, and Methacrylic Acid on SeQuant® ZIC-HILIC
HPLC Analysis of Methacrylamide, Acrylamide, and Methacrylic Acid on SeQuant® ZIC-HILIC application for HPLC
CONDITIONS
column
SeQuant ZIC-HILIC, PEEK, 150 x 4.6 mm, 5 μm, 200 Å (1.50455.0001)
mobile phase
[A] acetonitrile; [B] acetic acid; (95:5, A,B)
flow rate
1.0 mL/min
pressure
8.0 MPa
detector
210 nm
injection
5 μL
Description
Legal Information
SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany