MilliporeSigma
HPLC Analysis of Finasteride Tablets on Chromolith® HighResolution RP-18 endcapped 50 x 4.6mm
HPLC Analysis of Finasteride Tablets on Chromolith® HighResolution RP-18 endcapped 50 x 4.6mm application for HPLC
CONDITIONS
sample preparation
Dissolve an accurately weighed quantity of USP Finasteride RS in Diluting solution, and dilute quantitatively, and stepwise if necessary, with Diluting solution to obtain a solution having a known concentration of about 100 μg per mL.
column
Chromolith HighResolution RP-18 endcapped, 50 x 4.6 mm (1.52021.0001)
mobile phase
[A] acetonitrle; [B] 2.5 mM phosphoric acid in Water; (40:60,:A:B)
flow rate
2.25 mL/min
pressure
1160 psi 80 Bar
column temp.
45 °C
detector
240 nm cell: 11 μL
injection
10 μL
sample
Finasteride RS 100 μg/mL (100 ppm) (Diluent: Acetonitrile and water 70:30 (v/v))
Description
Legal Information
Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany