MilliporeSigma
HPLC Analysis of Benzoic Acid Derivatives on Ascentis® C18
HPLC Analysis of Benzoic Acid Derivatives on Ascentis® C18 application for HPLC
Materials
Analytical column
Product No.
Description
Pricing
Ascentis® C18 HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
Standard
Product No.
Description
Pricing
Acetylsalicylic acid
analytical standard
4-Hydroxybenzoic acid
ReagentPlus®, ≥99%
Benzoic acid
analytical standard
Ethylparaben
Pharmaceutical Secondary Standard; Certified Reference Material
CONDITIONS
column
Ascentis C18, 15 cm x 4.6 mm I.D., 5 μm particles (581324-U)
mobile phase
[A] 10mM TFA in water; [B] 10 mM TFA in acetonitrile; (70:30, A:B)
flow rate
1.0 mL/min
column temp.
30 °C
detector
UV, 220 nm
injection
10 μL
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany