US FDA LIB 4423: GC Analysis of Melamine and Related Compounds on SLB®-5ms, Method Blank
CONDITIONS
column
SLB-5ms, 30 m x 0.25 mm I.D., 0.25 μm (28471-U)
oven
115 °C (3 min), 10 °C/min to 325 °C (6 min)
inj. temp.
250 °C
MSD interface
325 °C
scan range
SIM
carrier gas
helium, 1 mL/min., constant
injection
1 μL splitless
liner
4 mm I.D. single taper
sample
TMS derivatives of melamine and related compounds (extract of a lab blank, with I.S. added at 1000 ng/mL, in pyridine:sylon BFT)
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany