GC Analysis of Fluorocarbons on a Packed Column (5% Fluorocol™ on 60/80 Carbopack™ B)
CONDITIONS
column
10’ x 1/8” SP® alloy, general configuration, packed with 5% Fluorcol™ on 60/80 Carbopack B
oven
50 °C
detector
FID
carrier gas
nitrogen, 30 mL/min
injection
10 μL synthetic mixture (approximately 20% each compound)
Description
Analysis Note
A packed GC column using Fluorcol stationary phase is also useful for fluorocarbons. The polarity of this column provides an alternate elution pattern compared to the Alumina chloride PLOT column chromatogram on the previous slide.
Legal Information
Carbopack is a trademark of Merck KGaA, Darmstadt, Germany
Fluorcol is a trademark of Sigma-Aldrich Co. LLC
SP is a registered trademark of Sigma-Aldrich Co. LLC