MilliporeSigma
  • Development of Pyridazinone Chemotypes Targeting the PDEδ Prenyl Binding Site.
Development of Pyridazinone Chemotypes Targeting the PDEδ Prenyl Binding Site.
Chemistry (Weinheim an der Bergstrasse, Germany) (2016-11-04)
Sandip Murarka, Pablo Martín-Gago, Carsten Schultz-Fademrecht, Alaa Al Saabi, Matthias Baumann, Eyad K Fansa, Shehab Ismail, Peter Nussbaumer, Alfred Wittinghofer, Herbert Waldmann
ABSTRACT
The K-Ras GTPase is a major target in anticancer drug discovery. However, direct interference with signaling by K-Ras has not led to clinically useful drugs yet. Correct localization and signaling by farnesylated K-Ras is regulated by the prenyl binding protein PDEδ. Interfering with binding of PDEδ to K-Ras by means of small molecules provides a novel opportunity to suppress oncogenic signaling. Here we describe the identification and structure-guided development of novel K-Ras-PDEδ inhibitor chemotypes based on pyrrolopyridazinones and pyrazolopyridazinones that bind to the farnesyl binding pocket of PDEδ with low nanomolar affinity. We delineate the structure-property relationship and in vivo pharmacokinetic (PK) and toxicokinetic (Tox) studies for pyrazolopyridazinone-based K-Ras-PDEδ inhibitors. These findings may inspire novel drug discovery efforts aimed at the development of drugs targeting oncogenic Ras.
Sigma-Aldrich
Deltarasin trihydrochloride, ≥98% (HPLC)