MilliporeSigma
  • Establishment of a three-step method to evaluate effects of chemicals on development of zebrafish embryo/larvae.
Establishment of a three-step method to evaluate effects of chemicals on development of zebrafish embryo/larvae.
Chemosphere (2017-08-07)
Guangyu Li, Huike Ye, Guanyong Su, Zhihua Han, Congxin Xie, Bingsheng Zhou, Robert J Letcher, John P Giesy, Hongxia Yu, Chunsheng Liu
ABSTRACT
Core endpoints in zebrafish embryos are crucial indicators in screening harmful effects of chemicals. In this study, we established a three-step process to more quantitatively and less-subjective determine effects of chemicals on phenotypes of developing zebrafish embryos. Embryos were exposed to each of two concentrations of the representative chemicals cadmium chloride (CdCl
Product Description
Supelco
TDCPP, PESTANAL®, analytical standard