MilliporeSigma
  • Metal based biologically active compounds: Design, synthesis, DNA binding and antidiabetic activity of 6-methyl-3-formyl chromone derived hydrazones and their metal (II) complexes.
Metal based biologically active compounds: Design, synthesis, DNA binding and antidiabetic activity of 6-methyl-3-formyl chromone derived hydrazones and their metal (II) complexes.
Journal of photochemistry and photobiology. B, Biology (2017-09-12)
Jessica Elizabeth Philip, Muhammad Shahid, M R Prathapachandra Kurup, Mohanan Puzhavoorparambil Velayudhan
ABSTRACT
Two chromone hydrazone ligands HL
Sigma-Aldrich
1,3-Di-o-tolylguanidine, 99%
Sigma-Aldrich
Benzhydrazide, 98%
Sigma-Aldrich
3-Formyl-6-methylchromone, 97%