MilliporeSigma
  • CYP17A1 inhibitor abiraterone, an anti-prostate cancer drug, also inhibits the 21-hydroxylase activity of CYP21A2.
CYP17A1 inhibitor abiraterone, an anti-prostate cancer drug, also inhibits the 21-hydroxylase activity of CYP21A2.
The Journal of steroid biochemistry and molecular biology (2017-09-13)
Jana Malikova, Simone Brixius-Anderko, Sameer S Udhane, Shaheena Parween, Bernhard Dick, Rita Bernhardt, Amit V Pandey
ABSTRACT
Abiraterone is an inhibitor of CYP17A1 which is used for the treatment of castration resistant prostate cancer. Abiraterone is known to inhibit several drug metabolizing cytochrome P450 enzymes including CYP1A2, CYP2D6, CYP2C8, CYP2C9, CYP2C19, CYP3A4 and CYP3A5, but its effects on steroid metabolizing P450 enzymes are not clear. In preliminary results, we had observed inhibition of CYP21A2 by 1μM abiraterone. Here we are reporting the effect of abiraterone on activities of CYP21A2 in human adrenal cells as well as with purified recombinant CYP21A2. Cells were treated with varying concentrations of abiraterone for 24h and CYP21A2 activity was measured using [
Product Description
Selenium, powder, 250 max. part. size (micron), purity 99.95%, weight 100 g