MilliporeSigma
  • Time-kill kinetic analysis of antimicrobial chemotherapy based on hydrogen peroxide photolysis against Streptococcus mutans biofilm.
Time-kill kinetic analysis of antimicrobial chemotherapy based on hydrogen peroxide photolysis against Streptococcus mutans biofilm.
Journal of photochemistry and photobiology. B, Biology (2017-07-01)
Midori Shirato, Keisuke Nakamura, Taro Kanno, Peter Lingström, Yoshimi Niwano, Ulf Örtengren
ABSTRACT
A recently developed antimicrobial technique utilizing hydroxyl radicals generated by hydrogen peroxide (H
Sigma-Aldrich
6,9-Diamino-2-ethoxyacridine-DL-lactate monohydrate, 99%