MilliporeSigma
  • Exposure to fine particulate matter in the air alters placental structure and the renin-angiotensin system.
Exposure to fine particulate matter in the air alters placental structure and the renin-angiotensin system.
PloS one (2017-08-19)
Sônia de Fátima Soto, Juliana Oliveira de Melo, Guilherme D'Aprile Marchesi, Karen Lucasechi Lopes, Mariana Matera Veras, Ivone Braga de Oliveira, Regiane Machado de Souza, Isac de Castro, Luzia Naôko Shinohara Furukawa, Paulo Hilário Nascimento Saldiva, Joel C Heimann
ABSTRACT
Female Wistar rats were exposed to filtered air (F) or to concentrated fine particulate matter (P) for 15 days. After mating, the rats were divided into four groups and again exposed to F or P (FF, FP, PF, PP) beginning on day 6 of pregnancy. At embryonic day 19, the placenta was collected. The placental structure, the protein and gene expression of TGFβ1, VEGF-A, and its receptor Flk-1 and RAS were evaluated by indirect ELISA and quantitative real-time PCR. Exposure to P decreased the placental mass, size, and surface area as well as the TGFβ1, VEGF-A and Flk-1 content. In the maternal portion of the placenta, angiotensin II (AngII) and its receptors AT1 (AT1R) and AT2 (AT2R) were decreased in the PF and PP groups. In the fetal portion of the placenta, AngII in the FP, PF and PP groups and AT2R in the PF and PP groups were decreased, but AT1R was increased in the FP group. VEGF-A gene expression was lower in the PP group than in the FF group. Exposure to pollutants before and/or during pregnancy alters some characteristics of the placenta, indicating a possible impairment of trophoblast invasion and placental angiogenesis with possible consequences for the maternal-fetal interaction, such as a limitation of fetal nutrition and growth.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution